This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07292999_u1
See other TRAPPC5 GE Assays ›
Gene Symbol:
TRAPPC5
Gene Name:
trafficking protein particle complex 5
Gene Aliases:
TRS31
Chromosome Location:
Chr.19: 7680821 - 7682862 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07292999_u1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042461.2 NP_001035926.1 2 216 72
NM_174894.2 NP_777554.1 2 95
GenBank mRNA BC042161.1 - 2 4869377 95
BI196928.1 - 2 159 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC042161.1, NM_001042461.2, NM_174894.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

Gene Ontology Categories:

Function(s) Process(es)

COPII vesicle coating
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w64qk:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0