| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
NM_000579_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
NM_001100168_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093303 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093300 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093302 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8079403 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8079404 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8079405 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093299 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8093301 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8079402 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8079406 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8079401 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8093298 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
206991_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11748062_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11730910_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11730909_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11730909_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11730910_PM_s_at |
- |
- |
| Affymetrix Human Genome U219 Array |
11748062_s_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
ENST00000343801.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
ENST00000400886.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
ENST00000445772.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005208.hg.1 |
TC03000256.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005209.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005209.hg.1 |
ENST00000343801.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005209.hg.1 |
ENST00000400886.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005209.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005209.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005209.hg.1 |
TC03000256.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
ENST00000445772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
ENST00000445772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151427.hg.1 |
TC0300007257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151430.hg.1 |
TC0300007257.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
ENST00000445772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
ENST00000292303.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
ENST00000445772.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
NM_000579.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
NM_001100168.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151429.hg.1 |
TC0300007257.hg.1 |
Clariom D TC |
| Human Genome U133 Plus 2.0 Array |
206991_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
206991_PM_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
206991_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
206991_PM_s_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw596078 |
- |
- |
| Human Genome U95 Set |
36724_PM_s_at |
- |
- |
| Human Genome U95 Set |
36724_s_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
206991_s_at1 |
- |
- |
| Human Genome U133 Set |
206991_s_at |
- |
- |
| Human Genome U133 Set |
206991_PM_s_at |
- |
- |