This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00559831_m1
See other EWSR1 GE Assays ›
Gene Symbol:
EWSR1
Gene Name:
EWS RNA binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 22205943 - 22239473 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00559831_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EWSR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287723.1 NP_001274652.1 3 - 4 116 70
XM_005567685.2 XP_005567742.1 3 - 4 425 70
XM_005567687.2 XP_005567744.1 3 - 4 425 70
XM_005567691.2 XP_005567748.1 3 - 4 420 70
XM_015456948.1 XP_015312434.1 3 - 4 420 70
XM_015456949.1 XP_015312435.1 3 - 4 420 70
XM_015456950.1 XP_015312436.1 3 - 4 420 70
XM_015456951.1 XP_015312437.1 3 - 4 420 70
XM_015456952.1 XP_015312438.1 3 - 4 420 70
GenBank mRNA AB179210.1 - 3 - 4 118 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Inventoried Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0