This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00608493_m1
See other ST18 GE Assays ›
Gene Symbol:
ST18
Gene Name:
ST18 C2H2C-type zinc finger transcription factor
Gene Aliases:
-
Chromosome Location:
Chr.8: 53070682 - 53398358 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00608493_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ST18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005563304.2 XP_005563361.1 21 - 22 5099 71
XM_015454842.1 XP_015310328.1 22 - 23 3330 71
XM_015454843.1 XP_015310329.1 22 - 23 3334 71
XM_015454844.1 XP_015310330.1 23 - 24 3487 71
XM_015454845.1 XP_015310331.1 21 - 22 3174 71
XM_015454846.1 XP_015310332.1 21 - 22 4974 71
XM_015454847.1 XP_015310333.1 21 - 22 3855 71
XM_015454848.1 XP_015310334.1 21 - 22 3251 71
XM_015454849.1 XP_015310335.1 21 - 22 3255 71
XM_015454850.1 XP_015310336.1 23 - 24 3501 71
XM_015454851.1 XP_015310337.1 20 - 21 5020 71
XM_015454852.1 XP_015310338.1 21 - 22 5191 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0