This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00900492_m1
See other NAPB GE Assays ›
Gene Symbol:
NAPB
Gene Name:
NSF attachment protein beta
Gene Aliases:
-
Chromosome Location:
Chr.10: 38839856 - 38891626 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00900492_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAPB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284026.1 NP_001270955.1 1 - 2 160 82
XM_015457631.1 XP_015313117.1 1 - 2 210 82
XM_015457632.1 XP_015313118.1 1 - 2 210 82
XM_015457633.1 XP_015313119.1 1 - 2 210 82
XM_015457634.1 XP_015313120.1 1 - 2 210 82
GenBank mRNA AB174308.1 - 1 - 2 160 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015457634.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0