This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00908917_g1
See other EIF4E GE Assays ›
Gene Symbol:
EIF4E
Gene Name:
eukaryotic translation initiation factor 4E
Gene Aliases:
-
Chromosome Location:
Chr.5: 98097195 - 98148343 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00908917_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4E
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005555502.2 XP_005555559.2 5 - 6 459 80
XM_005555503.2 XP_005555560.1 4 - 5 432 80
XM_005555504.2 XP_005555561.1 4 - 5 463 80
XM_005555505.2 XP_005555562.1 4 - 5 1808 80
GenBank mRNA AB168785.1 - 4 - 5 386 80
AB170765.1 - 4 - 5 294 80
AB171689.1 - 4 - 5 303 80
AB172042.1 - 829 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0