This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00920850_m1
See other TMEM218 GE Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
-
Chromosome Location:
Chr.14: 120908772 - 120925925 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00920850_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM218
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005580063.2 XP_005580120.1 3 - 4 490 89
XM_005580064.2 XP_005580121.1 3 - 4 530 89
XM_015435892.1 XP_015291378.1 3 - 4 475 89
XM_015435893.1 XP_015291379.1 3 - 4 465 89
XM_015435894.1 XP_015291380.1 3 - 4 331 89
XM_015435895.1 XP_015291381.1 3 - 4 808 89
XM_015435896.1 XP_015291382.1 3 - 4 480 89
XM_015435897.1 XP_015291383.1 3 - 4 520 89
XM_015435898.1 XP_015291384.1 3 - 4 321 89
XM_015435899.1 XP_015291385.1 3 - 4 510 89
XM_015435900.1 XP_015291386.1 3 - 4 525 89
XM_015435901.1 XP_015291387.1 2 - 3 896 89
XM_015435902.1 XP_015291388.1 3 - 4 462 89
XM_015435903.1 XP_015291389.1 2 - 3 935 89
XM_015435904.1 XP_015291390.1 3 - 4 564 89
XM_015435905.1 XP_015291391.1 3 - 4 477 89
XM_015435906.1 XP_015291392.1 3 - 4 516 89
XM_015435907.1 XP_015291393.1 3 - 4 317 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0