This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00950837_m1
See other ZCCHC17 GE Assays ›
Gene Symbol:
ZCCHC17
Gene Name:
zinc finger CCHC-type containing 17
Gene Aliases:
PS1D, pNO40
Chromosome Location:
Chr.1: 196669242 - 196739310 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00950837_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZCCHC17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285117.1 NP_001272046.1 5 - 6 419 87
XM_015452289.1 XP_015307775.1 5 - 6 602 87
XM_015452298.1 XP_015307784.1 5 - 6 462 87
XM_015452300.1 XP_015307786.1 4 - 5 493 87
GenBank mRNA AB060897.1 - 4 - 5 500 87
AB170145.1 - 5 - 6 419 87
AB171302.1 - 5 - 6 464 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0