This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00960672_g1
See other UBE2D3 GE Assays ›
Gene Symbol:
UBE2D3
Gene Name:
ubiquitin conjugating enzyme E2 D3
Gene Aliases:
-
Chromosome Location:
Chr.5: 102116805 - 102189795 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00960672_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2D3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283089.1 NP_001270018.1 6 - 7 585 76
XM_015450542.1 XP_015306028.1 6 - 7 798 76
XM_015450543.1 XP_015306029.1 6 - 7 664 76
XM_015450544.1 XP_015306030.1 6 - 7 741 76
XM_015450545.1 XP_015306031.1 6 - 7 783 76
XM_015450546.1 XP_015306032.1 6 - 7 713 76
XM_015450547.1 XP_015306033.1 6 - 7 957 76
XM_015450548.1 XP_015306034.1 6 - 7 664 76
XM_015450549.1 XP_015306035.1 6 - 7 792 76
XM_015450550.1 XP_015306036.1 6 - 7 713 76
XM_015450551.1 XP_015306037.1 5 - 6 306 76
XM_015450552.1 XP_015306038.1 5 - 6 459 76
GenBank mRNA AB169509.1 - 6 - 7 504 76
AB171036.1 - 6 - 7 585 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Inventoried Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline