This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00963440_g1
See other PTPA GE Assays ›
Gene Symbol:
PTPA
Gene Name:
protein phosphatase 2 phosphatase activator
Gene Aliases:
-
Chromosome Location:
Chr.15: 9992775 - 10033192 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00963440_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005580660.2 XP_005580717.1 4 - 5 625 57
XM_005580661.2 XP_005580718.1 3 - 4 538 57
XM_005580663.2 XP_005580720.1 4 - 5 625 57
XM_005580664.2 XP_005580721.1 3 - 4 538 57
GenBank mRNA AB170140.1 - 4 - 5 549 57
AB170536.1 - 4 - 5 548 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein-binding activity modulator phosphatase activator phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0