Assay Details
| Gene Symbol: RBM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283426.1 | NP_001270355.1 | 4 - 5 | 1454 | 57 | |
| XM_015434170.1 | XP_015289656.1 | 3 - 4 | 1519 | 57 | ||
| XM_015434171.1 | XP_015289657.1 | 3 - 4 | 1250 | 57 | ||
| XM_015434173.1 | XP_015289659.1 | 3 - 4 | 1790 | 57 | ||
| GenBank mRNA | AB168217.1 | - | 4 - 5 | 1454 | 57 | |
More Information
Set Membership: |
|

RBM4
Genomic Map