This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01046863_m1
See other GMDS GE Assays ›
Gene Symbol:
GMDS
Gene Name:
GDP-mannose 4,6-dehydratase
Gene Aliases:
-
Chromosome Location:
Chr.4: 168819121 - 169446627 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01046863_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GMDS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284855.1 NP_001271784.1 3 - 4 314 78
XM_005554174.2 XP_005554231.1 3 - 4 457 78
XM_015448795.1 XP_015304281.1 4 - 5 3284 78
XM_015448796.1 XP_015304282.1 2 - 3 1309 78
GenBank mRNA AB170853.1 - 3 - 4 314 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015448795.1, XM_015448796.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline