This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01065383_m1
See other GABPB1 GE Assays ›
Gene Symbol:
GABPB1
Gene Name:
GA binding protein transcription factor subunit beta 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 27253509 - 27330144 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01065383_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GABPB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283845.1 NP_001270774.1 4 - 5 645 124
XM_005559531.2 XP_005559588.1 4 - 5 1221 124
XM_005559535.2 XP_005559592.1 5 - 6 1245 124
XM_015452437.1 XP_015307923.1 4 - 5 924 124
XM_015452438.1 XP_015307924.1 5 - 6 1245 124
XM_015452439.1 XP_015307925.1 4 - 5 702 124
XM_015452440.1 XP_015307926.1 4 - 5 1221 124
XM_015452441.1 XP_015307927.1 4 - 5 1221 124
XM_015452442.1 XP_015307928.1 5 - 6 1245 124
XM_015452443.1 XP_015307929.1 3 - 4 1113 124
XM_015452444.1 XP_015307930.1 3 - 4 1113 124
GenBank mRNA AB174350.1 - 3 - 4 537 124
AB179213.1 - 4 - 5 645 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0