This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01116962_g1
See other NAT9 GE Assays ›
Gene Symbol:
NAT9
Gene Name:
N-acetyltransferase 9 (putative)
Gene Aliases:
-
Chromosome Location:
Chr.16: 72016137 - 72022283 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01116962_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005584874.2 XP_005584931.1 3 - 4 623 73
XM_005584875.2 XP_005584932.1 3 - 4 618 73
XM_005584876.2 XP_005584933.1 3 - 4 623 73
XM_005584877.2 XP_005584934.1 3 - 4 617 73
XM_005584878.2 XP_005584935.1 4 - 5 343 73
XM_005584879.2 XP_005584936.1 4 - 5 297 73
XM_005584880.2 XP_005584937.1 4 - 5 340 73
XM_015438809.1 XP_015294295.1 3 - 4 498 73
XM_015438810.1 XP_015294296.1 4 - 5 260 73
XM_015438811.1 XP_015294297.1 4 - 5 341 73
XM_015438812.1 XP_015294298.1 3 - 4 626 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0