This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01547652_m1
See other GLT8D2 GE Assays ›
Gene Symbol:
GLT8D2
Gene Name:
glycosyltransferase 8 domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 107674344 - 107752758 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01547652_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLT8D2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284696.1 NP_001271625.1 4 - 5 389 107
XM_005572056.2 XP_005572113.1 5 - 6 793 107
XM_005572058.2 XP_005572115.1 4 - 5 520 107
XM_005572059.2 XP_005572116.1 2 - 3 394 107
XM_015430643.1 XP_015286129.1 4 - 5 1250 107
XM_015430644.1 XP_015286130.1 5 - 6 483 107
XM_015430645.1 XP_015286131.1 4 - 5 409 107
XM_015430646.1 XP_015286132.1 4 - 5 373 107
XM_015430647.1 XP_015286133.1 4 - 5 745 107
GenBank mRNA AB179168.1 - 4 - 5 389 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0