This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01558089_m1
See other NAA60 GE Assays ›
Gene Symbol:
NAA60
Gene Name:
N-alpha-acetyltransferase 60, NatF catalytic subunit
Gene Aliases:
-
Chromosome Location:
Chr.20: 3697008 - 3739771 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01558089_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285386.1 NP_001272315.1 7 - 8 944 98
XM_005591065.2 XP_005591122.1 5 - 6 774 98
XM_015442141.1 XP_015297627.1 7 - 8 931 98
XM_015442143.1 XP_015297629.1 6 - 7 803 98
XM_015442144.1 XP_015297630.1 6 - 7 795 98
XM_015442145.1 XP_015297631.1 5 - 6 1117 98
XM_015442146.1 XP_015297632.1 5 - 6 594 98
XM_015442147.1 XP_015297633.1 4 - 5 646 98
GenBank mRNA AB171850.1 - 7 - 8 944 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0