This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf01572764_g1
See other OGFOD2 GE Assays ›
Gene Symbol:
OGFOD2
Gene Name:
2-oxoglutarate and iron dependent oxygenase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 127242993 - 127248563 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf01572764_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGFOD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005572523.2 XP_005572580.1 6 - 7 1446 81
XM_005572524.2 XP_005572581.1 5 - 6 1263 81
XM_005572525.2 XP_005572582.1 6 - 7 1393 81
XM_015431733.1 XP_015287219.1 6 - 7 1376 81
XM_015431734.1 XP_015287220.1 6 - 7 1323 81
XM_015431735.1 XP_015287221.1 3 - 4 450 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0