Assay Details
| Gene Symbol: LOC102120556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584243.2 | XP_005584300.1 | 10 - 11 | 1999 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102120556| Gene Symbol: LOC102120556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584243.2 | XP_005584300.1 | 10 - 11 | 1999 | 54 | |
Set Membership: |
|