This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02788385_m1
See other NR1I3 GE Assays ›
Gene Symbol:
NR1I3
Gene Name:
nuclear receptor subfamily 1 group I member 3
Gene Aliases:
CAR
Chromosome Location:
Chr.1: 90408741 - 90421652 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02788385_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NR1I3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319542.1 NP_001306471.1 8 - 9 1004 56
XM_005541186.2 XP_005541243.1 9 - 10 1579 56
XM_015454557.1 XP_015310043.1 8 - 9 1232 56
XM_015454567.1 XP_015310053.1 8 - 9 1217 56
XM_015454570.1 XP_015310056.1 8 - 9 1205 56
XM_015454575.1 XP_015310061.1 7 - 8 1103 56
XM_015454577.1 XP_015310063.1 7 - 8 1088 56
GenBank mRNA FJ202013.1 - 8 - 9 989 56
FJ202014.1 - 8 - 9 1004 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0