This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02792019_m1
See other ZSCAN32 GE Assays ›
Gene Symbol:
ZSCAN32
Gene Name:
zinc finger and SCAN domain containing 32
Gene Aliases:
-
Chromosome Location:
Chr.20: 3640420 - 3658951 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02792019_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZSCAN32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005591070.2 XP_005591127.1 6 - 7 1548 86
XM_015442293.1 XP_015297779.1 5 - 6 2661 86
XM_015442294.1 XP_015297780.1 4 - 5 1287 86
XM_015442295.1 XP_015297781.1 3 - 4 1075 86
XM_015442296.1 XP_015297782.1 4 - 5 2543 86
XM_015442297.1 XP_015297783.1 4 - 5 913 86
XM_015442298.1 XP_015297784.1 3 - 4 817 86
XM_015442299.1 XP_015297785.1 4 - 5 1062 86
XM_015442300.1 XP_015297786.1 3 - 4 714 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline