This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02797322_m1
See other METTL9 GE Assays ›
Gene Symbol:
METTL9
Gene Name:
methyltransferase like 9
Gene Aliases:
-
Chromosome Location:
Chr.20: 21307521 - 21360847 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02797322_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005591448.2 XP_005591505.1 3 - 4 818 91
XM_005591449.2 XP_005591506.1 3 - 4 818 91
XM_005591450.2 XP_005591507.1 3 - 4 818 91
XM_015442542.1 XP_015298028.1 3 - 4 818 91
XM_015442544.1 XP_015298030.1 3 - 4 480 91
XM_015442545.1 XP_015298031.1 3 - 4 480 91
XM_015442546.1 XP_015298032.1 3 - 4 419 91
GenBank mRNA AB171249.1 - 3 - 4 680 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline