This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02797762_m1
See other SDR16C5 GE Assays ›
Gene Symbol:
SDR16C5
Gene Name:
short chain dehydrogenase/reductase family 16C member 5
Gene Aliases:
-
Chromosome Location:
Chr.8: 57370174 - 57405513 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02797762_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDR16C5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283663.1 NP_001270592.1 2 - 3 845 67
XM_015454789.1 XP_015310275.1 3 - 4 1406 67
XM_015454790.1 XP_015310276.1 3 - 4 1657 67
XM_015454791.1 XP_015310277.1 2 - 3 602 67
GenBank mRNA AB174407.1 - 2 - 3 845 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bp22n:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0