This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02799167_m1
See other ESRRG GE Assays ›
Gene Symbol:
ESRRG
Gene Name:
estrogen related receptor gamma
Gene Aliases:
-
Chromosome Location:
Chr.1: 75127833 - 75778008 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02799167_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESRRG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005540851.2 XP_005540908.1 6 - 7 1966 127
XM_005540852.2 XP_005540909.1 6 - 7 9843 127
XM_005540853.2 XP_005540910.1 6 - 7 9818 127
XM_005540855.2 XP_005540912.1 7 - 8 1377 127
XM_005540856.2 XP_005540913.1 7 - 8 2931 127
XM_005540858.2 XP_005540915.1 7 - 8 1262 127
XM_005540859.2 XP_005540916.1 8 - 9 10137 127
XM_005540861.2 XP_005540918.1 7 - 8 1354 127
XM_005540862.2 XP_005540919.1 8 - 9 10111 127
XM_015450090.1 XP_015305576.1 7 - 8 10578 127
XM_015450091.1 XP_015305577.1 8 - 9 1426 127
XM_015450095.1 XP_015305581.1 6 - 7 2214 127
XM_015450096.1 XP_015305582.1 6 - 7 1621 127
XM_015450098.1 XP_015305584.1 7 - 8 1669 127
XM_015450101.1 XP_015305587.1 6 - 7 1169 127
GenBank mRNA AB172154.1 - 8 - 9 1425 127
AB174632.1 - 7 - 8 1754 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0