This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02809959_g1
See other EEF1D GE Assays ›
Gene Symbol:
EEF1D
Gene Name:
eukaryotic translation elongation factor 1 delta
Gene Aliases:
EF-1-delta, QtsA-17735
Chromosome Location:
Chr.8: 145214358 - 145233265 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02809959_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284982.1 NP_001271911.1 3 - 4 1362 89
XM_015454756.1 XP_015310242.1 3 - 4 3528 89
XM_015454757.1 XP_015310243.1 2 - 3 1256 89
XM_015454759.1 XP_015310245.1 4 - 5 1556 89
XM_015454760.1 XP_015310246.1 3 - 4 1662 89
XM_015454761.1 XP_015310247.1 2 - 3 443 89
XM_015454762.1 XP_015310248.1 2 - 3 508 89
GenBank mRNA AB169168.1 - 3 - 4 1362 89
AB179332.1 - 3 - 4 1362 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translation elongation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0