This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02812068_s1
See other IDI1 GE Assays ›
Gene Symbol:
IDI1
Gene Name:
Isopentenyl-diphosphate Delta-isomerase 1
Gene Aliases:
IPPI1
Chromosome Location:
Chr.9: 1223747 - 1252627 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02812068_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IDI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005564515.2 XP_005564572.1 2 613 137
XM_005564516.2 XP_005564573.1 2 452 137
XM_005564517.2 XP_005564574.1 2 613 137
GenBank mRNA AB169779.1 - 2 137 137
AB170542.1 - 2 89 137

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005564516.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

isomerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline