Assay Details
| Gene Symbol: HOGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566129.2 | XP_005566186.1 | 4 - 5 | 952 | 105 | |
| XM_005566130.2 | XP_005566187.1 | 4 - 5 | 967 | 105 | ||
More Information
Set Membership: |
|

HOGA1
Genomic Map