This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02826511_m1
See other CETP GE Assays ›
Gene Symbol:
CETP
Gene Name:
cholesteryl ester transfer protein
Gene Aliases:
-
Chromosome Location:
Chr.20: 44982595 - 45009714 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02826511_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CETP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005592006.2 XP_005592063.1 9 - 10 1512 92
XM_005592007.2 XP_005592064.1 10 - 11 1072 92
XM_005592008.2 XP_005592065.1 9 - 10 991 92
GenBank mRNA AB168680.1 - 9 - 10 1001 92
M86343.1 - 9 - 10 930 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0