This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02828383_g1
See other GALE GE Assays ›
Gene Symbol:
GALE
Gene Name:
UDP-galactose-4-epimerase
Gene Aliases:
-
Chromosome Location:
Chr.1: 204600497 - 204606457 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02828383_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GALE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319426.1 NP_001306355.1 9 - 10 970 109
XM_005544464.2 XP_005544521.1 10 - 11 1485 109
XM_015440860.1 XP_015296346.1 8 - 9 1222 109
XM_015440863.1 XP_015296349.1 9 - 10 1595 109
XM_015440867.1 XP_015296353.1 9 - 10 1108 109
XM_015440873.1 XP_015296359.1 8 - 9 1883 109
XM_015440877.1 XP_015296363.1 9 - 10 1595 109
XM_015440883.1 XP_015296369.1 9 - 10 1314 109
XM_015440888.1 XP_015296374.1 8 - 9 1503 109
XM_015440893.1 XP_015296379.1 9 - 10 1393 109
GenBank mRNA AB169339.1 - 9 - 10 970 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

epimerase/racemase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0