This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02836998_m1
See other HAGH GE Assays ›
Gene Symbol:
HAGH
Gene Name:
hydroxyacylglutathione hydrolase
Gene Aliases:
QtsA-18362
Chromosome Location:
Chr.20: 1864115 - 1881503 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02836998_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAGH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283414.1 NP_001270343.1 7 - 8 793 102
XM_005590906.2 XP_005590963.1 8 - 9 1251 102
XM_015442162.1 XP_015297648.1 8 - 9 1236 102
XM_015442164.1 XP_015297650.1 9 - 10 1283 102
XM_015442165.1 XP_015297651.1 6 - 7 983 102
GenBank mRNA AB169264.1 - 8 - 9 890 102
AB173495.1 - 7 - 8 795 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0