This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02837160_m1
See other PTPN20 GE Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase non-receptor type 20
Gene Aliases:
-
Chromosome Location:
Chr.9: 46922953 - 47024019 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02837160_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005565184.2 XP_005565241.1 8 - 9 882 104
XM_005565188.2 XP_005565245.1 7 - 8 736 104
XM_005565189.2 XP_005565246.1 7 - 8 1704 104
XM_005565193.2 XP_005565250.1 3 - 4 343 104
XM_005565194.2 XP_005565251.1 5 - 6 559 104
XM_015456058.1 XP_015311544.1 9 - 10 1157 104
XM_015456059.1 XP_015311545.1 8 - 9 1119 104
XM_015456060.1 XP_015311546.1 8 - 9 883 104
XM_015456061.1 XP_015311547.1 8 - 9 924 104
XM_015456062.1 XP_015311548.1 7 - 8 969 104
GenBank mRNA AB168280.1 - 6 - 7 697 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0