This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02851902_m1
See other PCMTD1 GE Assays ›
Gene Symbol:
PCMTD1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 52778493 - 52861269 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02851902_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMTD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005563298.2 XP_005563355.1 4 - 5 930 141
XM_005563299.2 XP_005563356.1 4 - 5 2574 141
XM_005563300.2 XP_005563357.1 3 - 4 505 141
XM_015454837.1 XP_015310323.1 4 - 5 702 141
XM_015454838.1 XP_015310324.1 4 - 5 930 141
XM_015454839.1 XP_015310325.1 4 - 5 720 141
XM_015454841.1 XP_015310327.1 2 - 3 370 141

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005563300.2, XM_015454839.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline