This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02864747_m1
See other SLCO3A1 GE Assays ›
Gene Symbol:
SLCO3A1
Gene Name:
solute carrier organic anion transporter family member 3A1
Gene Aliases:
-
Chromosome Location:
Chr.7: 72411270 - 72731061 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02864747_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO3A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005560546.2 XP_005560603.1 3 - 4 854 124
XM_005560547.2 XP_005560604.1 3 - 4 854 124
XM_005560548.2 XP_005560605.1 3 - 4 854 124
XM_005560549.2 XP_005560606.1 3 - 4 854 124
XM_005560550.2 XP_005560607.1 3 - 4 854 124
XM_005560551.2 XP_005560608.1 3 - 4 854 124
XM_015453421.1 XP_015308907.1 3 - 4 854 124

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0