Assay Details
| Gene Symbol: EIF2B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285349.1 | NP_001272278.1 | 2 - 3 | 275 | 75 | |
| XM_005543559.2 | XP_005543616.1 | 2 - 3 | 380 | 75 | ||
| XM_015451901.1 | XP_015307387.1 | 2 - 3 | 178 | 75 | ||
| GenBank mRNA | AB169097.1 | - | 2 - 3 | 263 | 75 | |
| AB170844.1 | - | 2 - 3 | 276 | 75 | ||
More Information
Set Membership: |
|

EIF2B3
Genomic Map