This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02879993_m1
See other STRA6 GE Assays ›
Gene Symbol:
STRA6
Gene Name:
signaling receptor and transporter of retinol STRA6
Gene Aliases:
-
Chromosome Location:
Chr.7: 52200379 - 52235132 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02879993_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STRA6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005560055.2 XP_005560112.1 3 - 4 366 149
XM_005560059.2 XP_005560116.1 3 - 4 344 149
XM_015453069.1 XP_015308555.1 3 - 4 473 149
XM_015453070.1 XP_015308556.1 4 - 5 448 149
XM_015453071.1 XP_015308557.1 3 - 4 328 149
XM_015453072.1 XP_015308558.1 3 - 4 426 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8t6tc:80/100.66.75.151:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0