This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02884309_g1
See other ARHGAP36 GE Assays ›
Gene Symbol:
ARHGAP36
Gene Name:
Rho GTPase activating protein 36
Gene Aliases:
-
Chromosome Location:
Chr.X: 127963909 - 127995550 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02884309_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP36
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005594576.1 XP_005594633.1 6 - 7 1086 120
XM_005594577.2 XP_005594634.1 7 - 8 1054 120
XM_005594578.2 XP_005594635.1 7 - 8 883 120
XM_005594579.2 XP_005594636.1 7 - 8 890 120
XM_015444310.1 XP_015299796.1 7 - 8 986 120
XM_015444311.1 XP_015299797.1 6 - 7 801 120
XM_015444312.1 XP_015299798.1 6 - 7 715 120

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline