This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02884517_m1
See other NXPE3 GE Assays ›
Gene Symbol:
NXPE3
Gene Name:
neurexophilin and PC-esterase domain family member 3
Gene Aliases:
-
Chromosome Location:
Chr.2: 170345578 - 170387384 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02884517_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005548256.2 XP_005548313.1 7 - 8 2152 117
XM_005548259.2 XP_005548316.1 6 - 7 1877 117
XM_015446507.1 XP_015301993.1 8 - 9 1939 117
XM_015446508.1 XP_015301994.1 7 - 8 2118 117
XM_015446509.1 XP_015301995.1 8 - 9 1913 117
XM_015446510.1 XP_015301996.1 7 - 8 1741 117

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0