Assay Details
| Gene Symbol: GPR156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548050.2 | XP_005548107.1 | 3 - 4 | 1356 | 144 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR156| Gene Symbol: GPR156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548050.2 | XP_005548107.1 | 3 - 4 | 1356 | 144 | |
Set Membership: |
|