Assay Details
| Gene Symbol: SLC25A41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587708.2 | XP_005587765.1 | 3 - 4 | 796 | 157 | |
| XM_005587709.2 | XP_005587766.1 | 3 - 4 | 406 | 157 | ||
More Information
Set Membership: |
|

SLC25A41
Genomic Map