This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02902834_m1
See other NAT1 GE Assays ›
Gene Symbol:
NAT1
Gene Name:
N-acetyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 18237091 - 18250652 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02902834_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005562697.2 XP_005562754.1 2 - 3 184 158
XM_005562699.2 XP_005562756.1 3 - 4 411 158
XM_015454519.1 XP_015310005.1 3 - 4 549 158
XM_015454520.1 XP_015310006.1 4 - 5 792 158
XM_015454521.1 XP_015310007.1 2 - 3 2111 158

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005562697.2, XM_005562699.2, XM_015454519.1, XM_015454520.1, XM_015454521.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Inventoried Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0