Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581044.2 | XP_005581101.1 | 4 | 2484 | 125 | |
| XM_015436854.1 | XP_015292340.1 | 4 | 2483 | 125 | ||
| XM_015436855.1 | XP_015292341.1 | 4 | 2354 | 125 | ||
More Information
Set Membership: |
|

ZFP37
Genomic Map