This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04344813_m1
See other COMT GE Assays ›
Gene Symbol:
COMT
Gene Name:
catechol-O-methyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.10: 31485200 - 31513714 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04344813_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005567961.2 XP_005568018.1 5 - 6 1377 60
XM_005567962.2 XP_005568019.1 5 - 6 867 60
XM_015457422.1 XP_015312908.1 6 - 7 1381 60
XM_015457423.1 XP_015312909.1 6 - 7 996 60
XM_015457424.1 XP_015312910.1 7 - 8 1033 60
XM_015457425.1 XP_015312911.1 5 - 6 1288 60
XM_015457426.1 XP_015312912.1 4 - 5 632 60
XM_015457427.1 XP_015312913.1 3 - 4 893 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0