This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04345060_m1
See other NUDT1 GE Assays ›
Gene Symbol:
NUDT1
Gene Name:
nudix hydrolase 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 37180640 - 37194090 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04345060_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005549007.2 XP_005549064.1 3 - 4 560 106
XM_005549008.2 XP_005549065.1 3 - 4 615 106
XM_005549010.2 XP_005549067.1 3 - 4 528 106
XM_005549012.2 XP_005549069.1 3 - 4 668 106
XM_015447021.1 XP_015302507.1 3 - 4 612 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0