This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04345307_g1
See other ELOVL1 GE Assays ›
Gene Symbol:
ELOVL1
Gene Name:
ELOVL fatty acid elongase 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 184251897 - 184256487 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04345307_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELOVL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283586.1 NP_001270515.1 4 - 5 416 90
XM_005543676.2 XP_005543733.1 4 - 5 1904 90
XM_005543677.2 XP_005543734.1 4 - 5 582 90
GenBank mRNA AB170918.1 - 4 - 5 416 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0