This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04346105_m1
See other HMOX2 GE Assays ›
Gene Symbol:
HMOX2
Gene Name:
heme oxygenase 2
Gene Aliases:
HO-2
Chromosome Location:
Chr.20: 4730483 - 4778583 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04346105_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HMOX2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283226.1 NP_001270155.1 4 - 5 785 92
XM_015442166.1 XP_015297652.1 4 - 5 875 92
XM_015442168.1 XP_015297654.1 5 - 6 1029 92
XM_015442169.1 XP_015297655.1 4 - 5 923 92
XM_015442170.1 XP_015297656.1 4 - 5 878 92
XM_015442171.1 XP_015297657.1 4 - 5 830 92
GenBank mRNA AB220384.1 - 4 - 5 785 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

oxygenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0