This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04355240_g1
See other METTL15 GE Assays ›
Gene Symbol:
METTL15
Gene Name:
methyltransferase like 15
Gene Aliases:
-
Chromosome Location:
Chr.14: 38937519 - 39377315 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04355240_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001284720.1 NP_001271649.1 6 - 7 1076 64
XM_015434089.1 XP_015289575.1 5 - 6 977 64
XM_015434090.1 XP_015289576.1 5 - 6 937 64
XM_015434091.1 XP_015289577.1 4 - 5 1369 64
XM_015434092.1 XP_015289578.1 5 - 6 955 64
XM_015434093.1 XP_015289579.1 5 - 6 955 64
XM_015434094.1 XP_015289580.1 5 - 6 955 64
GenBank mRNA AB168804.1 - 6 - 7 1076 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline