This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04360858_m1
See other MUTYH GE Assays ›
Gene Symbol:
MUTYH
Gene Name:
mutY DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.1: 182191295 - 182203761 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04360858_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MUTYH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283675.1 NP_001270604.1 14 - 15 1457 101
XM_015451760.1 XP_015307246.1 15 - 16 1700 101
XM_015451766.1 XP_015307252.1 15 - 16 1697 101
XM_015451769.1 XP_015307255.1 15 - 16 1588 101
XM_015451773.1 XP_015307259.1 15 - 16 1554 101
XM_015451777.1 XP_015307263.1 15 - 16 2145 101
XM_015451779.1 XP_015307265.1 14 - 15 1621 101
XM_015451781.1 XP_015307267.1 15 - 16 1594 101
XM_015451787.1 XP_015307273.1 14 - 15 1471 101
XM_015451790.1 XP_015307276.1 15 - 16 1702 101
GenBank mRNA AB179425.1 - 14 - 15 1457 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline