This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04373640_g1
See other HSD11B1L GE Assays ›
Gene Symbol:
HSD11B1L
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases:
11-DH3, HSD3
Chromosome Location:
Chr.19: 5813651 - 5824772 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04373640_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005587623.2 XP_005587680.1 6 - 7 1131 107
XM_005587625.2 XP_005587682.1 6 - 7 1113 107
XM_005587626.2 XP_005587683.1 6 - 7 1092 107
XM_005587627.2 XP_005587684.1 6 - 7 1077 107
XM_005587628.2 XP_005587685.1 6 - 7 1074 107
XM_005587634.2 XP_005587691.1 7 - 8 1029 107
XM_005587635.2 XP_005587692.1 7 - 8 991 107
XM_005587636.1 XP_005587693.1 7 - 8 1192 107
XM_005587639.1 XP_005587696.1 6 - 7 1129 107
XM_005587640.1 XP_005587697.1 6 - 7 1080 107
XM_005587641.1 XP_005587698.1 5 - 6 1017 107
XM_015440269.1 XP_015295755.1 7 - 8 952 107
XM_015440270.1 XP_015295756.1 7 - 8 990 107
XM_015440271.1 XP_015295757.1 7 - 8 969 107
XM_015440273.1 XP_015295759.1 4 - 5 1045 107
XM_015440274.1 XP_015295760.1 6 - 7 818 107
GenBank mRNA AB046006.1 - 7 - 8 932 107
AY554048.1 - 7 - 8 897 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0