Assay Details
| Gene Symbol: SIGIRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576725.2 | XP_005576782.1 | 5 - 6 | 1063 | 60 | |
| GenBank mRNA | AB179231.1 | - | 5 - 6 | 785 | 60 | |
More Information
Set Membership: |
|

SIGIRR
Genomic Map