This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04382840_m1
See other CDH18 GE Assays ›
Gene Symbol:
CDH18
Gene Name:
cadherin 18
Gene Aliases:
-
Chromosome Location:
Chr.6: 19781734 - 20916154 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04382840_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005556617.2 XP_005556674.1 8 - 9 1686 88
XM_015451230.1 XP_015306716.1 7 - 8 1610 88
XM_015451231.1 XP_015306717.1 8 - 9 1829 88
XM_015451232.1 XP_015306718.1 8 - 9 1825 88
XM_015451234.1 XP_015306720.1 7 - 8 1591 88
XM_015451235.1 XP_015306721.1 7 - 8 1393 88
XM_015451236.1 XP_015306722.1 6 - 7 1790 88
XM_015451237.1 XP_015306723.1 7 - 8 1610 88
XM_015451238.1 XP_015306724.1 7 - 8 1610 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0