This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf04382847_g1
See other POLR3G GE Assays ›
Gene Symbol:
POLR3G
Gene Name:
RNA polymerase III subunit G
Gene Aliases:
-
Chromosome Location:
Chr.6: 88878651 - 88920099 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf04382847_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005557353.2 XP_005557410.1 2 - 3 312 86
XM_005557354.2 XP_005557411.1 2 - 3 274 86
XM_005557355.2 XP_005557412.1 2 - 3 271 86
XM_005557356.2 XP_005557413.1 2 - 3 257 86
XM_005557357.2 XP_005557414.1 2 - 3 285 86
XM_005557358.2 XP_005557415.1 2 - 3 229 86
XM_015451684.1 XP_015307170.1 2 - 3 286 86
XM_015451685.1 XP_015307171.1 1 - 2 1934 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0